3,5-Bis(trifluoromethyl)phenylboronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5-Bis(trifluoromethyl)benzeneboronic acid, (3,5-Bis(trifluoromethyl)phenyl)metaboric acid, (3,5-Bis(trifluoromethyl)phenyl)boronic acid, Dihydroxy(3,5-bis(trifluoromethyl)phenyl)borane

35136-0010 35136-0050
TS35136-0010EA 33 USD
TS35136-0010 TS35136-0050
3,5-Bis(trifluoromethyl)phenylboronic acid 95%
3,5-Bis(trifluoromethyl)phenylboronic acid
Formula: C₈H₅BF₆O₂
Melting Pt: 219…226 °C
MDL Number: MFCD00051850
CAS Number: 73852-19-4

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Melting point 219 to 226 °C
Titration with NaOH 94 to 108 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR