2,4,6-Trimethylbenzylamine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Mesitylmethanamine, 1-Mesitylmethanamine
35118-0010
35118-0050
TS35118-0010EA
37
USD
TS35118-0010
TS35118-0050
2,4,6-Trimethylbenzylamine 99%
2,4,6-Trimethylbenzylamine
Formula:
C₁₀H₁₅N MW: 149.24 |
MDL Number:
MFCD03411013 CAS Number: 40393-99-5 UN: 2735 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5360 to 1.5380 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...