(4-Chlorophenyl)acetic acid methyl ester 99%

Supplier: Thermo Scientific Chemicals
Warning

35105-0050 35105-0250
TS35105-0050EA 51 USD
TS35105-0050 TS35105-0250
(4-Chlorophenyl)acetic acid methyl ester 99%
(4-Chlorophenyl)acetic acid methyl ester
Formula: (C₆H₄Cl)CH₂COOCH₃
MDL Number: MFCD00032743
CAS Number: 52449-43-1
EINECS: 257-927-0

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5220 to 1.5240 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR