(2-Chlorophenyl)acetic acid methyl ester 98%

Supplier: Thermo Scientific Chemicals
Warning

35104-0050 35104-0250
TS35104-0050EA 47 USD
TS35104-0050 TS35104-0250
(2-Chlorophenyl)acetic acid methyl ester 98%
(2-Chlorophenyl)acetic acid methyl ester
Formula: (C₆H₄Cl)CH₂COOCH₃
MDL Number: MFCD00137445
CAS Number: 57486-68-7
EINECS: 260-764-8

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5240 to 1.5260 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR