2,6-Dichlorobenzylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

35074-0050 35074-0250
TS35074-0050EA 60 USD
TS35074-0050 TS35074-0250
2,6-Dichlorobenzylamine 98%
2,6-Dichlorobenzylamine
Formula: C₇H₇Cl₂N
MDL Number: MFCD00047928
CAS Number: 6575-27-5
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5770 to 1.5790 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR