(4-Chlorophenyl)acetone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Chlorophenylacetone

35070-0050 35070-0250
TS35070-0050EA 96 USD
TS35070-0050 TS35070-0250
(4-Chlorophenyl)acetone 98%
(4-Chlorophenyl)acetone
Formula: C₉H₉ClO
MW: 168.62 g/mol
Boiling Pt: 132 °C (12 mmHg)
Density: 1.151 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00045214
CAS Number: 5586-88-9
EINECS: 226-986-4

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5325 to 1.5345 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR