(3-Chlorophenyl)acetone 94%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(3-Chlorophenyl)propan-2-one

35069-0010 35069-0050 35069-0250
TS35069-0010EA 28 USD
TS35069-0010 TS35069-0050 TS35069-0250
(3-Chlorophenyl)acetone 94%
(3-Chlorophenyl)acetone
Formula: C₉H₉ClO
MW: 168.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082872
CAS Number: 14123-60-5

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5320 to 1.5360 (20°C, 589 nm)
GC ≥92 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR