(3-Bromophenyl)acetone 98+%

Supplier: Thermo Scientific Chemicals

35060-0050 35060-0250
TS35060-0050EA 116 USD
TS35060-0050 TS35060-0250
(3-Bromophenyl)acetone 98+%
(3-Bromophenyl)acetone
Formula: C₉H₉BrO
Storage Temperature: Ambient
MDL Number: MFCD00210400
CAS Number: 21906-32-1

Order Now

Specification Test Results

Appearance Clear light yellow to dark yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5570 to 1.5600 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR