4-Bromobenzylamine 97+%

Supplier: Thermo Scientific Chemicals
Danger

35053-0050 35053-0250
TS35053-0050EA 205 USD
TS35053-0050 TS35053-0250
4-Bromobenzylamine 97+%
4-Bromobenzylamine
Formula: C₇H₈BrN
MW: 186.05 g/mol
Melting Pt: 25 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00047931
CAS Number: 3959-07-7
EINECS: 223-559-4
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid or solid
Infrared spectrum Conforms
Refractive index 1.5860 to 1.5890 (20°C, 589 nm)
GC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR