2-Bromobenzylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (2-Bromophenyl)methanamine

35051-0050 35051-0250
TS35051-0050EA 75 USD
TS35051-0050 TS35051-0250
2-Bromobenzylamine 98%
2-Bromobenzylamine
Formula: C₇H₈BrN
MW: 186.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00025572
CAS Number: 3959-05-5
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5880 to 1.5930 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR