(S)-(-)-2-(Diphenylmethyl)pyrrolidine 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-2-Diphenylmethylpyrrolidine

34837-0010 34837-0050 34837-2500
TS34837-0010EA 127 USD
TS34837-0010 TS34837-0050 TS34837-2500
(S)-(-)-2-(Diphenylmethyl)pyrrolidine 97+%
(S)-(-)-2-(Diphenylmethyl)pyrrolidine
Formula: C₁₇H₁₉N
MW: 237.35 g/mol
Density: 1.062 g/cm³
Flash Pt: 109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00799525
CAS Number: 119237-64-8

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5860 to 1.5880 (20°C, 589 nm)
GC ≥97.0 %
Specific optical rotation -2° to -4° (25°C, 589 nm) (c=1, chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR