6-Hydroxy-2-naphthoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-hydroxynaphthalene-2-carboxylic acid

34765-0050 34765-0250
TS34765-0050EA 58 USD
TS34765-0050 TS34765-0250
6-Hydroxy-2-naphthoic acid 98%
6-Hydroxy-2-naphthoic acid
Formula: C₁₁H₈O₃
MW: 188.18 g/mol
Melting Pt: 240…250 °C
MDL Number: MFCD00060070
CAS Number: 16712-64-4
EINECS: 240-759-7

Order Now

Specification Test Results

Appearance Light brown Powder
Infrared spectrum Conforms
Melting point 240 to 250 °C
Titration with TBAH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR