4-Bromobenzyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Bromo-α-chlorotoluene, α-Chloro-4-bromotoluene, 1-Bromo-4-(chloromethyl)benzene, 1-(Chloromethyl)-4-bromobenzene

34675-0050 34675-0250
TS34675-0050EA 52 USD
TS34675-0050 TS34675-0250
4-Bromobenzyl chloride 98%
4-Bromobenzyl chloride
Formula: C₇H₆BrCl
Boiling Pt: 136…139 °C (27 mmHg)
Melting Pt: 36…40 °C
MDL Number: MFCD00040867
CAS Number: 589-17-3
EINECS: 209-638-6
UN: 3261
ADR: 8,II
Merck Index: 13,01394

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 36 to 40 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR