2,4,5-Trihydroxybenzaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

34594-0010 34594-0050
TS34594-0010EA 160 USD
TS34594-0010 TS34594-0050
2,4,5-Trihydroxybenzaldehyde 99%
2,4,5-Trihydroxybenzaldehyde
Formula: C₇H₆O₄
MW: 154.12 g/mol
Melting Pt: 226…227 °C
MDL Number: MFCD00016592
CAS Number: 35094-87-2

Order Now

Specification Test Results

Appearance Yellow to brown to khaki Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR