3-Fluorophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

34564-0010 34564-0050 34564-0250
TS34564-0010EA 45 USD
TS34564-0010 TS34564-0050 TS34564-0250
3-Fluorophenylboronic acid 97%
Formula: C₆H₆BFO₂
Melting Pt: 211…212 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236042
CAS Number: 768-35-4

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR