4-tert-Butyl-1-(bromomethyl)benzene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-(Bromomethyl)-4-(tert-butyl)benzene, 4-(tert-Butyl)benzylbromide

34550-0050 34550-0250 34550-1000
TS34550-0050EA 37 USD
TS34550-0050 TS34550-0250 TS34550-1000
4-tert-Butyl-1-(bromomethyl)benzene 97%
4-tert-Butyl-1-(bromomethyl)benzene
Formula: C₁₁H₁₅Br
MW: 227.14 g/mol
Boiling Pt: 90…94 °C (1.5 mmHg)
Melting Pt: 8…12 °C
Density: 1.236 g/cm³
Flash Pt: >109 °C
Storage Temperature: Ambient
MDL Number: MFCD00000180
CAS Number: 18880-00-7
EINECS: 242-643-1
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5445 to 1.5465 (20°C, 589 nm)
Titration Argentometric ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR