4-tert-Butyl-α-chlorotoluene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: p-tert-Butylbenzyl chloride, 4-tert-Butylbenzyl chloride, p-tert-Butyl-α-chlorotoluene, 1-(tert-Butyl)-4-(chloromethyl)benzene

34549-0050 34549-0250
TS34549-0050EA 58 USD
TS34549-0050 TS34549-0250
4-tert-Butyl-α-chlorotoluene 97%
4-tert-Butyl-α-chlorotoluene
Formula: C₁₁H₁₅Cl
MW: 182.69 g/mol
Boiling Pt: 98…103 °C (7 mmHg)
Density: 0.94 g/cm³
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00000918
CAS Number: 19692-45-6
EINECS: 243-228-8
UN: 1760
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5205 to 1.5225 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR