N-Ethylbenzylamine 95%

Supplier: Thermo Scientific Chemicals
Danger

34371-0250 34371-1000 34371-5000
TS34371-0250EA 40 USD
TS34371-0250 TS34371-1000 TS34371-5000
N-Ethylbenzylamine 95%
N-Ethylbenzylamine
Formula: C₆H₅CH₂NHCH₂CH₃
Boiling Pt: 191…194 °C (760 mmHg)
Density: 0.9 g/cm³
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00009031
CAS Number: 14321-27-8
EINECS: 238-265-1
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5100 to 1.5130 (20°C, 589 nm)
Titration with HClO4 ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR