α-Bromo-4-iodotoulene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Iodobenzyl bromide, 1-(Bromomethyl)-4-iodobenzene, α-Bromo-p-iodotoulene

34246-0010 34246-0050 34246-0250
TS34246-0010EA 31 USD
TS34246-0010 TS34246-0050 TS34246-0250
α-Bromo-4-iodotoulene 97%
α-Bromo-4-iodotoulene
Formula: C₇H₆BrI
MW: 296.93
Boiling Pt: 140 °C (15 mmHg)
Melting Pt: 80
Storage Temperature: Refrigerator
MDL Number: MFCD00209656
CAS Number: 16004-15-2
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Melting point 77 to 82 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR