Benzophenone oxime 98%

Supplier: Thermo Scientific Chemicals
Danger

34233-0050 34233-0250
TS34233-0050EA 37 USD
TS34233-0050 TS34233-0250
Benzophenone oxime 98%
Benzophenone oxime
Formula: C₁₃H₁₁NO
Melting Pt: 140…144 °C
Storage Temperature: Freezer
MDL Number: MFCD00051461
CAS Number: 574-66-3
EINECS: 209-373-6

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 140 to 144 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR