5-Methoxy-2-nitrophenol 98%

Supplier: Thermo Scientific Chemicals
Danger

33544-0010 33544-0050
TS33544-0010EA 130 USD
TS33544-0010 TS33544-0050
5-Methoxy-2-nitrophenol 98%
5-Methoxy-2-nitrophenol
Formula: C₇H₇NO₄
Melting Pt: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00100932
CAS Number: 704-14-3

Order Now

Specification Test Results

Appearance Yellow Powder or crystals
Infrared spectrum Conforms
Melting point 90 to 94 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR