4-Amino-3-nitrophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxy-2-nitroaniline

33522-0050 33522-0250 33522-1000
TS33522-0050EA 32 USD
TS33522-0050 TS33522-0250 TS33522-1000
4-Amino-3-nitrophenol 98%
4-Amino-3-nitrophenol
Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00066310
CAS Number: 610-81-1
EINECS: 210-236-8

Order Now

Specification Test Results

Appearance Red to bordeaux Crystalline powder
Infrared spectrum Conforms
Melting point ≥150 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR