1,2-Benzenedithiol 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzene-1,2-dithiol

33492-0010 33492-0050
TS33492-0010EA 143 USD
TS33492-0010 TS33492-0050
1,2-Benzenedithiol 96%
1,2-Benzenedithiol
Formula: C₆H₆S₂
MW: 142.25 g/mol
Melting Pt: 27…28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004835
CAS Number: 17534-15-5
EINECS: 241-530-4

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid or solid
Infrared spectrum Conforms
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR