2-Bromo-5-nitroaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

33486-0050 33486-0250
TS33486-0050EA 56 USD
TS33486-0050 TS33486-0250
2-Bromo-5-nitroaniline 98%
2-Bromo-5-nitroaniline
Formula: C₆H₅BrN₂O₂
Melting Pt: 138…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00051578
CAS Number: 10403-47-1
EINECS: 233-874-9

Order Now

Specification Test Results

Appearance Yellow to yellow-green to yellow-brown Powder or crystals or needles
Infrared spectrum Conforms
Melting point 137 to 142 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR