4,6-Diaminoresorcinol dihydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,6-Dihydroxy-1,3-phenylenediamine Dihydrochloride, 4,6-Diaminobenzene-1,3-diol dihydrochloride

33367-0050 33367-0250 33367-1000
TS33367-0050EA 95 USD
TS33367-0050 TS33367-0250 TS33367-1000
4,6-Diaminoresorcinol dihydrochloride 98%
4,6-Diaminoresorcinol dihydrochloride
Formula: C₆H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00143239
CAS Number: 16523-31-2

Order Now

Specification Test Results

Appearance Grey-brown Crystalline solid
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR