1-Bromo-3,5-dichlorobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5-Dichlorobromobenzene

33190-0050 33190-0250
TS33190-0050EA 18 USD
TS33190-0050 TS33190-0250 TS33190-1000
1-Bromo-3,5-dichlorobenzene 98%
1-Bromo-3,5-dichlorobenzene
Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 232 °C
Melting Pt: 74…77 °C
Flash Pt: >110 °C
MDL Number: MFCD00000584
CAS Number: 19752-55-7
EINECS: 243-270-7

Order Now

Specification Test Results

Appearance White to beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 74 to 77 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR