6-Bromo-2-tetralone 98%

Supplier: Thermo Scientific Chemicals

33077-0010 33077-0050
TS33077-0010EA 55 USD
TS33077-0010 TS33077-0050
6-Bromo-2-tetralone 98%
6-Bromo-2-tetralone
Formula: C₁₀H₉BrO
Melting Pt: 70…76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00239388
CAS Number: 4133-35-1

Order Now

Specification Test Results

Appearance Light yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 70 to 76 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR