3-Iodo-4-nitroanisole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Iodo-4-methoxy-1-nitrobenzene

32632-0050 32632-0250
TS32632-0050EA 87 USD
TS32632-0050 TS32632-0250
3-Iodo-4-nitroanisole 97%
3-Iodo-4-nitroanisole
Formula: C₇H₆INO₃
Melting Pt: 68…73 °C
Storage Temperature: Ambient
MDL Number: MFCD01320682
CAS Number: 214279-40-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to orange-brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 68 to 73 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR