Ethyl-3,4-dihydroxybenzoate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,4-Dihydroxybenzoic acid ethyl ester, Protocatechuic acid ethyl ester, Ethyl 3,4-dihydroxybenzoate
32581-0050
32581-0250
TS32581-0050EA
37
USD
TS32581-0050
TS32581-0250
Ethyl-3,4-dihydroxybenzoate 97%
Ethyl-3,4-dihydroxybenzoate
Formula:
C₉H₁₀O₄ MW: 182.18 g/mol Melting Pt: 131…136 °C |
MDL Number:
MFCD00002199 CAS Number: 3943-89-3 EINECS: 223-529-0 |
Specification Test Results
Appearance | Light yellow to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 131 to 136 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...