1-Bromo-4-butylbenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

32044-0100 32044-0500
TS32044-0100EA 32 USD
TS32044-0100 TS32044-0500
1-Bromo-4-butylbenzene 97%
1-Bromo-4-butylbenzene
Formula: C₁₀H₁₃Br
Density: 1.2 g/cm³
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00040934
CAS Number: 41492-05-1

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5300 to 1.5330 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR