(1S,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol 99%

Supplier: Thermo Scientific Chemicals
Warning

31565-0010 31565-0050 31565-2500
TS31565-0010EA 49 USD
TS31565-0010 TS31565-0050 TS31565-2500
(1S,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol 99%
(1S,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol
Formula: C₉H₁₁NO
Melting Pt: 117…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00216655
CAS Number: 126456-43-7

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 117 to 121 °C
GC ≥98.5 %
Specific optical rotation -41.5° to -46.5° (20 °C, 589 nm) (c=1, CH3OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR