2-(Benzyloxy)ethanol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethylene Glycol Monobenzyl Ether, Benzyl Cellosolve, Benzyl Glycol
31465-0010
31465-0050
31465-0250
TS31465-0010EA
29
USD
TS31465-0010
TS31465-0050
TS31465-0250
2-(Benzyloxy)ethanol 98%
2-(Benzyloxy)ethanol
Formula:
C₉H₁₂O₂ MW: 152.19 g/mol Boiling Pt: 265 °C Density: 1.07 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002868 CAS Number: 622-08-2 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5200 to 1.5220 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...