4-(Chloromethyl)benzoyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

31444-0050 31444-0250
TS31444-0050EA 57 USD
TS31444-0050 TS31444-0250
4-(Chloromethyl)benzoyl chloride 97%
4-(Chloromethyl)benzoyl chloride
Formula: C₈H₆Cl₂O
Boiling Pt: 126…128 °C (6 mmHg)
Melting Pt: 29…33 °C
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00053224
CAS Number: 876-08-4
EINECS: 212-881-0
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to light beige Low melting crystalline mass
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR