3-(Chloromethyl)benzoyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
31443-0050
31443-0250
TS31443-0050EA
61
USD
TS31443-0050
TS31443-0250
3-(Chloromethyl)benzoyl chloride 98%
3-(Chloromethyl)benzoyl chloride
Formula:
C₈H₆Cl₂O MW: 189.04 g/mol Boiling Pt: 149…150 °C (20 mmHg) Melting Pt: 4 °C Density: 1.33 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000682 CAS Number: 63024-77-1 EINECS: 263-799-7 UN: 3265 ADR: 8,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5740 to 1.5760 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...