α'-Bromo-α,α,α-trifluoro-p-xylene 98%

Supplier: Thermo Scientific Chemicals
Danger

31299-0010 31299-0050 31299-0250
TS31299-0010EA 32 USD
TS31299-0010 TS31299-0050 TS31299-0250
α'-Bromo-α,α,α-trifluoro-p-xylene 98%
α'-Bromo-α,α,α-trifluoro-p-xylene
Formula: C₈H₆BrF₃
Boiling Pt: 65…69 °C (5 mmHg)
Melting Pt: 30…34 °C
Density: 1.54 g/cm³
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00000403
CAS Number: 402-49-3
EINECS: 206-947-8
UN: 1760
ADR: 8,III

Order Now

Specification Test Results

Appearance White to light yellow Low melting crystalline solid
Infrared spectrum Conforms
Melting point 30 to 34 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR