Pentafluoroiodobenzene 99% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Iodopentafluorobenzene

Stabilization: Stabilised with copper

31220-0010 31220-0050
TS31220-0010EA 21 USD
TS31220-0010 TS31220-0050
Pentafluoroiodobenzene 99% stabilized
Pentafluoroiodobenzene
Formula: C₆F₅I
MW: 293.96 g/mol
Boiling Pt: 162…165 °C (760 mmHg)
Density: 2.204 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00001032
CAS Number: 827-15-6

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR