2,6-Difluorophenol 98%

Supplier: Thermo Scientific Chemicals
Danger

31163-0250 31163-0050
TS31163-0250EA 254 USD
TS31163-0250 TS31163-0050
2,6-Difluorophenol 98%
2,6-Difluorophenol
Formula: C₆H₄F₂O
MW: 130.09 g/mol
Boiling Pt: 59…61 °C (17 mmHg)
Melting Pt: 38…42 °C
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00002158
CAS Number: 28177-48-2
EINECS: 248-884-9
UN: 2925
ADR: 4.1,III

Order Now

Specification Test Results

Appearance Colorless Low melting crystalline solid
Infrared spectrum Conforms
Melting point 38 to 42 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR