3,5-Difluoroaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

31142-0010 31142-0100 31142-0500
TS31142-0010EA 29 USD
TS31142-0010 TS31142-0100 TS31142-0500
3,5-Difluoroaniline 98%
3,5-Difluoroaniline
Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 82 °C (20 mmHg)
Melting Pt: 37…41 °C
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00007763
CAS Number: 372-39-4
EINECS: 206-752-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow Low melting solid (May darken on storage)
Infrared spectrum Conforms
Melting point 37 to 41 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR