1,2-Dibromotetrafluorobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-Dibromo-3,4,5,6-tetrafluorobenzene

31135-0010 31135-0050
TS31135-0010EA 54 USD
TS31135-0010 TS31135-0050
1,2-Dibromotetrafluorobenzene 99%
1,2-Dibromotetrafluorobenzene
Formula: C₆Br₂F₄
MW: 307.87 g/mol
Boiling Pt: 198 °C
Melting Pt: 12
Density: 2.23 g/cm³
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00039209
CAS Number: 827-08-7
EINECS: 212-564-7

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5145 to 1.5175 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR