Ethyl-3-bromobenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 3-bromobenzoate, 3-Bromobenzoic acid ethyl ester

30854-0050 30854-0250
TS30854-0050EA 21 USD
TS30854-0050 TS30854-0250
Ethyl-3-bromobenzoate 98%
Ethyl-3-bromobenzoate
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 130…131 °C (12 mmHg)
Density: 1.43 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00013529
CAS Number: 24398-88-7
EINECS: 246-223-9

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5405 to 1.5425 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR