3,5-Bis(trifluoromethyl)benzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

30769-0010 30769-0050 30769-0250
TS30769-0010EA 27 USD
TS30769-0010 TS30769-0050 TS30769-0250
3,5-Bis(trifluoromethyl)benzaldehyde 97%
3,5-Bis(trifluoromethyl)benzaldehyde
Formula: C₉H₄F₆O
MW: 242.12 g/mol
Boiling Pt: 37 °C (1.3 mmHg)
Density: 1.47 g/cm³
Flash Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD00010206
CAS Number: 401-95-6

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4215 to 1.4235 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR