3-Bromostyrene 97% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-3-vinylbenzene

30648-0010 30648-0050
TS30648-0010EA 44 USD
TS30648-0010 TS30648-0050
3-Bromostyrene 97% stabilized
3-Bromostyrene
Formula: C₈H₇Br
MW: 183.05 g/mol
Boiling Pt: 74…75 °C (3 mmHg)
Density: 1.406 g/cm³
Flash Pt: 67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000088
CAS Number: 2039-86-3
EINECS: 218-025-2

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
Stabilizer 0.05 % 3,5-di-tert.-Butylcatechol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR