3,5-Bis(trifluoromethyl)benzylamine 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (3,5-Bis(trifluoromethyl)phenyl)methanamine

30564-0010 30564-0050
TS30564-0010EA 46 USD
TS30564-0010 TS30564-0050
3,5-Bis(trifluoromethyl)benzylamine 95%
3,5-Bis(trifluoromethyl)benzylamine
Formula: C₉H₇F₆N
MW: 243.15 g/mol
Boiling Pt: 82…84 °C (15 mmHg)
Melting Pt: 46…55 °C
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00009909
CAS Number: 85068-29-7
EINECS: 285-290-9
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals or mass and/or chunks
Infrared spectrum Conforms
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR