2,3-Dimethylhydroquinone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,4-Dihydroxy-2,3-dimethylbenzene

30511-0050 30511-0250 30511-1000
TS30511-0050EA 96 USD
TS30511-0050 TS30511-0250 TS30511-1000
2,3-Dimethylhydroquinone 97%
2,3-Dimethylhydroquinone
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Melting Pt: 223…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00009997
CAS Number: 608-43-5

Order Now

Specification Test Results

Appearance Off-white to light brown to purple Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR