3,5-Bis(trifluoromethyl)benzyl bromide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-(Bromomethyl)-3,5-bis(trifluoromethyl)benzene
30364-0010
30364-0050
TS30364-0010EA
42
USD
TS30364-0010
TS30364-0050
3,5-Bis(trifluoromethyl)benzyl bromide 97%
3,5-Bis(trifluoromethyl)benzyl bromide
Formula:
C₉H₅BrF₆ MW: 307.03 g/mol Boiling Pt: 136…140 °C (14 mmHg) Density: 1.67 g/cm³ Flash Pt: 26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009905 CAS Number: 32247-96-4 EINECS: 250-971-1 UN: 2920 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow-brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4440 to 1.4460 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...