3-Fluoro-4-nitrophenol 99%

Supplier: Thermo Scientific Chemicals
Warning

30337-0010 30337-0050
TS30337-0010EA 35 USD
TS30337-0010 TS30337-0050
3-Fluoro-4-nitrophenol 99%
3-Fluoro-4-nitrophenol
Formula: C₆H₄FNO₃
Melting Pt: 92…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00041251
CAS Number: 394-41-2
EINECS: 206-895-6
UN: 2811

Order Now

Specification Test Results

Appearance Light yellow to brown Powder
Infrared spectrum Conforms
Melting point 91 to 95 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR