4-Chloro-3-sulfamoylbenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

30322-0010 30322-0050
TS30322-0010EA 41 USD
TS30322-0010 TS30322-0050
4-Chloro-3-sulfamoylbenzoic acid 98%
4-Chloro-3-sulphamoylbenzoic acid
Formula: C₇H₆ClNO₄S
Boiling Pt: 256…258 °C (760 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00012375
CAS Number: 1205-30-7

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 258 to 263 °C
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR