1,2,3,4-Tetrahydronaphthalen-2-one (2-tetralone-β-tetralone) 95%

Supplier: Thermo Scientific Chemicals
Warning

30283-0050 30283-0250
TS30283-0050EA 56 USD
TS30283-0050 TS30283-0250
1,2,3,4-Tetrahydronaphthalen-2-one (2-tetralone-β-tetralone) 95%
1,2,3,4-Tetrahydronaphthalen-2-one (2-tetralone-β-tetralone)
Formula: C₁₀H₁₀O
Boiling Pt: 131…133 °C (11 mmHg)
Melting Pt: 18 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001727
CAS Number: 530-93-8
EINECS: 208-498-3

Order Now

Specification Test Results

Appearance Clear colorless to yellow to green Liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.5550 to 1.5650 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR