Bis(phenylsulfonyl)sulfide 96%

Supplier: Thermo Scientific Chemicals

Synonyms: Benzenesulphonic thioanhydride

30083-0010 30083-0050
TS30083-0010EA 85 USD
TS30083-0010 TS30083-0050
Bis(phenylsulfonyl)sulfide 96%
Bis(phenylsulfonyl)sulfide
Formula: C₁₂H₁₀O₄S₃
MW: 314.388 g/mol
Melting Pt: 129…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00094733
CAS Number: 4388-22-1

Order Now

Specification Test Results

Appearance Off-white to pink Powder or crystals
Infrared spectrum Conforms
Melting point 129 to 134 °C
HPLC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR