6-Methoxy-2-tetralone 90%
Supplier: Thermo Scientific Chemicals
|
Warning
|
29892-0010
29892-0050
TS29892-0010EA
113
USD
TS29892-0010
TS29892-0050
6-Methoxy-2-tetralone 90%
6-Methoxy-2-tetralone
Formula:
C₁₁H₁₂O₂ Boiling Pt: 114…116 °C (.2 mmHg) Melting Pt: 28…35 °C Flash Pt: >110 °C Storage Temperature: Freezer |
MDL Number:
MFCD00001729 CAS Number: 2472-22-2 EINECS: 219-592-9 |
Specification Test Results
Appearance | Yellow to light beige to orange Low melting crystalline solid |
Infrared spectrum | Conforms |
Melting point | 28 to 35 °C |
GC | ≥88 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...